missing translation for 'onlineSavingsMsg'
Learn More

Aluminum perchlorate nonahydrate, Reagent Grade

Catalog No. p-7048851
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1165922 100 g
AA1165936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1165922 Supplier Thermo Scientific Chemicals Supplier No. 01165922
Only null left

CAS: 81029-06-3 | AlCl3H18O21 | 487.455 g/mol

Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 81029-06-3
Molecular Formula AlCl3H18O21
Molecular Weight (g/mol) 487.455
MDL Number MFCD00149130
InChI Key CYRDFMGMNQJULY-UHFFFAOYSA-K
Synonym aluminum perchlorate nonahydrate, al.3clo4.9h2o, aluminum perchlorate nonahydrate, reagent, aluminum 3+ nonahydrate triperchlorate
PubChem CID 16211522
IUPAC Name aluminum;triperchlorate;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Al+3]

Specifications

Melting Point 82°C
Physical Form Crystalline
Quantity 100 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 487.46 (325.33 Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Aluminum perchlorate nonahydrate
Hazard Category H272-H314
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 238-437-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only