accessibility menu, dialog, popup

UserName

CAS RN 81029-06-3

H2OH2OH2OH2OH2OH2OH2OH2OH2OOClOOOOClOOOOClOOOAl

CAS RN 81029-06-3

IUPAC Name: aluminium(3+) nonahydrate triperchlorate
Synonyms: aluminium(3+) nonahydrate triperchlorate
Molecular Weight (g/mol): 487.45
Molecular Formula: AlCl3H18O21
InChi Key: CYRDFMGMNQJULY-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.O.O.O.[Al+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results