Learn More
Aluminum Nitrate Nonahydrate (Cryst./Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLA5863
Specifications
| Aluminum Nitrate Nonahydrate | |
| 7784-27-2 | |
| Solid | |
| 98.0 to 102.0 % | |
| Al(NO3)3·9H2O | |
| aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
| 375.129 | |
| 375.13 | |
| Certified ACS | |
| Glass Bottle |
| 73°C | |
| 2.5 to 3.5 | |
| 3 kg | |
| AlH18N3O18 | |
| MFCD00149132 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate | |
| 24567 | |
| 98.0 to 102.0% | |
| Pass Test |
Chemical Identifiers
| 7784-27-2 | |
| 375.129 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| 24567 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| AlH18N3O18 | |
| MFCD00149132 | |
| aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
| aluminum;trinitrate;nonahydrate |
Safety and Handling
DANGER!
Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:1
Instability:2
DOTInformation : DOT Class 5.1, : Oxidizer