missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Nitrate Nonahydrate (Cryst./Certified ACS), Fisher Chemical™

Catalog No. p-30769
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A586500 500 g Glass Bottle
A5863 3 kg Glass Bottle
A58610 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A586500 Supplier Thermo Fisher Scientific Supplier No. FLA586500
Only null left

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00149132
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73°C
pH 2.5 to 3.5
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0 %
Linear Formula Al(NO3)3·9H2O
Formula Weight 375.13
Percent Purity 98.0 to 102.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Aluminum Nitrate Nonahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Hygroscopic. Use personal protective equipment. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer