Learn More
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Supplier: Thermo Scientific Chemicals 03629130
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
| Aluminum nitrate nonahydrate | |
| nonahydrate | |
| White | |
| 250 g | |
| AlH18N3O18 | |
| aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate | |
| 24567 | |
| ACS | |
| Hygroscopic |
| 73.5°C (135° (decomposition)) | |
| 7784-27-2 | |
| Crystalline | |
| 98 -102% | |
| MFCD00003419 | |
| Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine. | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
| 375.129 | |
| 375.13 (212.99 Anhydrous) | |
| Odorless | |
| White crystalline |
Chemical Identifiers
| 7784-27-2 | |
| 375.129 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| 24567 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| AlH18N3O18 | |
| MFCD00003419 | |
| aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
| aluminum;trinitrate;nonahydrate |
Safety and Handling
P280-P305+P351+P338-P310
H318
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 236-751-8
RTECSNumber : BD1050000
TSCA : Yes
storageNote1 : Hygroscopic
Recommended Storage : Ambient temperatures
RUO – Research Use Only