missing translation for 'onlineSavingsMsg'
Learn More

Aluminum nitrate nonahydrate, ACS, 98.0-102.0%

Catalog No. p-3256856
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
5 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3629118 50 g
AA3629130 250 g
AA36291A1 1 kg
AA36291D9 5 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA3629118 Supplier Thermo Scientific Chemicals Supplier No. 03629118
Only null left

CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 7784-27-2
Molecular Formula AlH18N3O18
Molecular Weight (g/mol) 375.129
MDL Number MFCD00003419
InChI Key SWCIQHXIXUMHKA-UHFFFAOYSA-N
Synonym aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic
PubChem CID 24567
IUPAC Name aluminum;trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3]

Specifications

Melting Point 73.5°C (135° (decomposition))
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 98 -102%
Solubility Information Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine.
Formula Weight 375.13 (212.99 Anhydrous)
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Aluminum nitrate nonahydrate, nonahydrate
Show More Show Less
Hazard Category H318
Precautionary Statement P280-P305+P351+P338-P310
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 236-751-8
RTECSNumber BD1050000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only