missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Hydroxide, Powder, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A109325KG

 View more versions of this product

Catalog No. 18608517

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

21645-51-2
2.5 kg
Reagent
1
95%
Poly Pail

Chemical Identifiers

21645-51-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.