missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Hydroxide, Powder, 95%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystalline Powder or Needles |
| Quantity | 500 g |
| Molecular Weight (g/mol) | 78.00 |
| Percent Purity | 95% |
| Grade | Reagent |
| Packaging | Poly Pail |
| Chemical Name or Material | Aluminum Hydroxide |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.