Learn More
Aluminum hydroxide, 76.5% min
CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol
$47.30 - $284.22
Chemical Identifiers
| CAS | 21645-51-2 |
|---|---|
| Molecular Formula | AlH3O3 |
| Molecular Weight (g/mol) | 78.00 |
| MDL Number | MFCD00003420 |
| InChI Key | WNROFYMDJYEPJX-UHFFFAOYSA-K |
| IUPAC Name | aluminium(3+) trihydroxide |
| SMILES | [OH-].[OH-].[OH-].[Al+3] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1236622
|
Thermo Scientific Chemicals
01236622 |
100 g |
Each for $47.30
|
|
|||||
|
AA1236636
|
Thermo Scientific Chemicals
01236636 |
500 g |
Each for $133.11
|
|
|||||
|
AA12366A3
|
Thermo Scientific Chemicals
012366A3 |
2 kg |
Each for $284.22
|
|
|||||
Description
As absorbent, in chromatography, manufacturing of glass, paper, inks, ceramics, lubricants, cosmeticsAluminum hydroxide is used as a desiccant powder, filler in paper, plastics, rubber and cosmetics. It is used as a smoke suppressant and mordant dye. It is also used in drugs as antacid and antihyperphosphatemic. It is also used as a Claus catalyst support for waterproof fabrics. It is an important starting material for the preparation of other aluminum compounds, calcined alumina, aluminum sulfate, polyaluminum chloride, zeolites, sodium aluminate, activated alumina and aluminum nitrate. In addition, it is used as a fire retardant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 21645-51-2 | |
| 78.00 | |
| WNROFYMDJYEPJX-UHFFFAOYSA-K | |
| [OH-].[OH-].[OH-].[Al+3] |
| AlH3O3 | |
| MFCD00003420 | |
| aluminium(3+) trihydroxide |
Specifications
| 21645-51-2 | |
| 100 g | |
| AlH3O3 | |
| Soluble in alkaline aqueous solution,hydrochloric acid and sulfuric acid solution. Insoluble in water. | |
| [OH-].[OH-].[OH-].[Al+3] | |
| 78.00 | |
| Reagent |
| Granules | |
| 76.5% min. | |
| MFCD00003420 | |
| WNROFYMDJYEPJX-UHFFFAOYSA-K | |
| aluminium(3+) trihydroxide | |
| 78 | |
| Aluminum hydroxide |
Safety and Handling
H315-H319-H335
EINECSNumber : 244-492-7
RTECSNumber : BD0940000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only