missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Glycinate 97.0+%, TCI America™
$82.02 - $361.62
Chemical Identifiers
| CAS | 13682-92-3 |
|---|---|
| Molecular Formula | C6H15AlN3O6 |
| Molecular Weight (g/mol) | 252.183 |
| MDL Number | MFCD00053645 |
| InChI Key | VBASKOIIBZSXOZ-UHFFFAOYSA-N |
| Synonym | Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate |
| PubChem CID | 129894307 |
| IUPAC Name | aluminum;2-aminoacetic acid |
| SMILES | C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.[Al] |
Chemical Identifiers
| 13682-92-3 | |
| 252.183 | |
| VBASKOIIBZSXOZ-UHFFFAOYSA-N | |
| 129894307 | |
| C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.[Al] |
| C6H15AlN3O6 | |
| MFCD00053645 | |
| Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate | |
| aluminum;2-aminoacetic acid |
Specifications
| 13682-92-3 | |
| C6H15AlN3O6 | |
| 25 g | |
| VBASKOIIBZSXOZ-UHFFFAOYSA-N | |
| aluminum;2-aminoacetic acid | |
| 129894307 | |
| ≥97.0% (T) | |
| Aluminum Glycinate |
| White | |
| MFCD00053645 | |
| Aminoacetic Acid Dihydroxyaluminum Salt, Dihydroxyaluminum Aminoacetate | |
| C(C(=O)O)N.C(C(=O)O)N.C(C(=O)O)N.[Al] | |
| 252.183 | |
| 135.05 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes