accessibility menu, dialog, popup

UserName

CAS RN 13682-92-3

OHOH2NOHOH2NOHOH2NAl

CAS RN 13682-92-3

IUPAC Name: tris(2-aminoacetic acid) aluminium
Synonyms: tris(glycine) aluminium
Molecular Weight (g/mol): 252.18
Molecular Formula: C6H15AlN3O6
InChi Key: VBASKOIIBZSXOZ-UHFFFAOYSA-N
SMILES: [Al].NCC(O)=O.NCC(O)=O.NCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results