missing translation for 'onlineSavingsMsg'
Learn More

Aluminum ethoxide

CAS: 555-75-9 | C6H15AlO3-3 | 162.165 g/mol

$1444.01 - $1444.01

Chemical Identifiers

CAS 555-75-9
Molecular Formula C6H15AlO3-3
Molecular Weight (g/mol) 162.165
MDL Number MFCD00009658
InChI Key JBRFPFQROJYWFN-UHFFFAOYSA-N
Synonym acmc-209lnt
PubChem CID 70700059
IUPAC Name aluminum;ethanolate
SMILES CC[O-].CC[O-].CC[O-].[Al]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4133630
View Documents
Thermo Scientific Chemicals
04133630
250 g
Each for $1,444.01
Only null left
 
Description

Description

Aluminum ethoxide is mainly used in industrial settings. It is used as a reducing agent for aldehyde, ketones, polymerization catalyst. Used in the sol-gel process preparation of high purity aluminum sesquioxide. It also used as a reducing reagent, for carbonyl compounds that restore to alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

555-75-9
162.165
JBRFPFQROJYWFN-UHFFFAOYSA-N
70700059
CC[O-].CC[O-].CC[O-].[Al]
C6H15AlO3-3
MFCD00009658
acmc-209lnt
aluminum;ethanolate

Specifications

555-75-9
White
320°C
Al(OC2H5)3
250 g
Moisture sensitive
acmc-209lnt
JBRFPFQROJYWFN-UHFFFAOYSA-N
aluminum;ethanolate
70700059
Chunks and/or Powder
154°C to 159°C
1.142 g/mL
C6H15AlO3-3
MFCD00009658
UN2925
14,338
Soluble in hot xylene,cholorobenzene. Reacts with water.
CC[O-].CC[O-].CC[O-].[Al]
162.165
162.16
Aluminum ethoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H314
Flammable solid.
Causes severe skin burns and eye damage.

P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H228-H261-H314

DOTInformation : Hazard Class: 4.3; Packaging Group: II

EINECSNumber : 209-105-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only