missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Aluminum Danger Sign: FLAMMABLE MATERIALS.
Improve workplace safety with proper signage and markings. This Brady™ corner hole mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
Supplier: Brady™ 145051
Description
- Aluminum material is abrasion-resistant and chemical-resistant
- Aluminum signs provide the strength and durability of metal, but never rust and can tolerate extreme temperatures, years of chemical spills, wash downs and cleanings
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor and outdoor use
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Chemical Identifiers
| Not Laminated | |
| 1 Ea. | |
| Corner Holes | |
| -40°F | |
| Rectangular | |
| FLAMMABLE MATERIALS. |
| Aluminum | |
| Non-adhesive | |
| 230°F | |
| 0.035 in. | |
| -40°F to 230°F |
Specifications
| Not Laminated | |
| 1 Ea. | |
| Corner Holes | |
| 230°F | |
| 0.035 in. | |
| Rectangular | |
| 14 x 10 in. | |
| Red |
| Aluminum | |
| Non-adhesive | |
| 10 in. | |
| -40°F | |
| 14 in. | |
| -40°F to 230°F | |
| FLAMMABLE MATERIALS. |