missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Aluminum crimp seal (without septum)

Catalog No. 111020959
Change view
Click to view available options
Diameter (Metric) Outer:
20 mm
Style:
Open Center
Pressure Release
Removable Center
Tear-away
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Style
111020959 20 mm Pressure Release
111020960 20 mm Tear-away
111016057 20 mm Removable Center
111012056 20 mm Open Center
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 111020959 Supplier MilliporeSigma™ Supelco™ Supplier No. SGMA27200
Only null left

Seal without septum used to seal 100 mL serum bottles

  • Used for sealing 100 mL serum bottles
  • Fitted with gray stopper during analysis of biogas emission in lagoons.

Specifications

Diameter (Metric) Opening 8 mm
Color Silver
Diameter (Metric) Outer 20 mm
Material Aluminum
No. per Pack 100
Style Pressure Release
For Use With (Equipment) 20 mm Crimp Seal Vial