missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Aluminum crimp seal (without septum)
Seal without septum used to seal 100 mL serum bottles
Supplier: MilliporeSigma™ Supelco™ 27230U
Description
- Used for sealing 100 mL serum bottles
- Fitted with gray stopper during analysis of biogas emission in lagoons.
Chemical Identifiers
| 100 |
Specifications
| 9.5 mm | |
| 20 mm | |
| 100 | |
| 20 mm Crimp Seal Vial |
| Silver | |
| Aluminum | |
| Open Center |