missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Copper Square bar, 1.27 x 1.27cm (0.5 x 0.5 in.), Thermo Scientific™

Form: Approx.4.48g/cm

Specifications

Chemical Name or Material Aluminum Copper
Molecular Formula Al:Cu:Mg:Mn; 93.5:4.4:1.5:0.6 wt%
MDL Number MFCD00213972
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42103BT
View Documents
Thermo Scientific Chemicals
42103BT
15 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA42103RS
View Documents
Thermo Scientific Chemicals
42103RS
60 cm N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum Copper
Al:Cu:Mg:Mn; 93.5:4.4:1.5:0.6 wt%
MFCD00213972
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'