missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Copper Square bar, 1.27 x 1.27cm (0.5 x 0.5 in.), Thermo Scientific™
Form: Approx.4.48g/cm
Supplier: Thermo Scientific Chemicals 42103RS
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Aluminum Copper | |
| 60 cm | |
| Al:Cu:Mg:Mn; 93.5:4.4:1.5:0.6 wt% |
| Silver-colored bar | |
| Odorless | |
| MFCD00213972 |