missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Chloride, Anhydrous, 95%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A107845KGBL

 View more versions of this product

Catalog No. 18608499


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7446-70-0
45 kg
Reagent
1
95%
Fiber Drum

Chemical Identifiers

7446-70-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.