missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Chloride, Anhydrous, 95%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 100 g |
| Percent Purity | 95% |
| Grade | Reagent |
| Packaging | Amber Glass Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.