missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum chloride, 6.0N Standardized Solution
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Physical Form | Liquid |
| Concentration | 6.0N Standardized Solution |
| UN Number | UN2581 |
| Type | Analytical Volumetric Solution |
| Quantity | 250 mL |
| Formula Weight | 133.34 |
| Grade | Standard |
| Chemical Name or Material | Aluminum chloride |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H StatementH314-H318Causes severe skin burns and eye damage.Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALUMINUM CHLORIDE, SOLUTION |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only