missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, 6.0N Standardized Solution

Catalog No. AA35568AE
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35568AE 100 mL
AA35568AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA35568AE Supplier Thermo Scientific Chemicals Supplier No. 035568AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7446-70-0 | AlCl3 | 133.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Odor Odorless
Density 1.3g/cm3 at 20°C
Physical Form Liquid
Color Colorless
Quantity 100 mL
Formula Weight 133.34
Chemical Name or Material Aluminum chloride
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packing Group: III
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only