Learn More
Troemner™ Aluminum ASTM Class 2 Weight with ISO/IEC 17025 Accredited Certificate
Troemener Individual Calibration Weights are Adjusted to Meet or Exceed ASTM Class 1-4 Tolerances
Supplier:  Troemner™ 80858705
Catalog No. 02218259
Only null left
Explore available promotions
Description
Description
  • Troemner ASTM Class 1-4 offer high accuracy with tight tolerances and uncertainties of less than 1/4 of the tolerance. Weight surfaces meet or exceed ASTM and have a highly polished finish.
  • Available in high grade stainless steels including Troemner Alloy 8 which possesses closely controlled density, extremely low magnetic properties, good stability, and resistance to corrosion.
  • Available with certificate options traceable to NIST including ISO/IEC 17025 Accredited Certificate or Traceable Certificate to aid in meeting SOP and regulatory requirements.
Specifications
Specifications
Calibration Mass
Aluminum
1 mg
Protective Case
ISO/IEC 17025 Accredited Certificate
Polycarbonate
Flat
Product Suggestions
Product Suggestions