missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Ammonium Sulfate, Dodecahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor A107525KG

 View more versions of this product

Catalog No. 18608495


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7784-26-1
2.5 kg
ACS
1
98 to 102%
Poly Bottle

Chemical Identifiers

7784-26-1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.