missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Ammonium Sulfate, Dodecahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. p-7164958
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608493 25 g Poly Bottle
18608494 500 g Poly Bottle
18608495 2.5 kg Poly Bottle
18608496 12 kg Poly Pail
18608497 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608493 Supplier Spectrum Chemical Mfg Cor Supplier No. A107525GM
Only null left

Chemical Identifiers

CAS 7784-26-1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98 to 102%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.