missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL

Catalog No. AA33557AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33557AE 100 mL
AA33557AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33557AE Supplier Thermo Scientific Chemicals Supplier No. 033557AE

CAS: 11145-27-0 | AlSi | 55.067 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 11145-27-0
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00134029
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Density 1g/cm3 at 20°C
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material Aluminum
Hazard Category H290
Precautionary Statement P234-P390
DOTInformation DOT Class: 8; Packing Group: III
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only