Learn More
Aluminum AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Al), LabChem™
Supplier: LabChem LC107501
Specifications
| Aluminum AA Standard | |
| Poly Bottle | |
| 95.51,3.6,0.89 | |
| Liquid | |
| 1000 ppm ±10 ppm (1mL = 1mg Al) | |
| UN1789 | |
| Soluble in water | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
| 375.129 | |
| 24567 | |
| Certified |
| 1000ppm (1mL = 1mg Al) | |
| 7784-27-2,7647-01-0,7784-13-6,7732-18-5 | |
| Passes Test | |
| AlH18N3O18 | |
| AlCl3·6H2O | |
| Colorless | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate | |
| 500 mL | |
| 241.43 |
Chemical Identifiers
| 7784-27-2 | |
| 375.129 | |
| 24567 | |
| [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
| AlH18N3O18 | |
| SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
| aluminum;trinitrate;nonahydrate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
Recommended Storage : Room Temperature