missing translation for 'onlineSavingsMsg'
Learn More

Aluminum 2,4-pentanedionate, 99.995+% (metals basis)

Catalog No. p-7059405
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4453806 5 g
AA4453814 25 g
AA4453822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4453806 Supplier Thermo Scientific Chemicals Supplier No. 04453806
Only null left

CAS: 13963-57-0 | C15H21AlO6 | 324.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym Aluminum acetylacetonate
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 192°C to 194°C
Density 1.27 g/mL
Boiling Point 314°C to 316°C
Quantity 5 g
UN Number UN3467
Beilstein 4157942
Formula Weight 324.31
Percent Purity ≥99.995%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Aluminum 2,4-pentanedionate
Show More Show Less
Hazard Category H300-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 237-741-6
RTECSNumber BD2230000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only