missing translation for 'onlineSavingsMsg'
Learn More

Aluminon, ACS reagent

Catalog No. p-4380036
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC401500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC401500250 Supplier Thermo Scientific Chemicals Supplier No. 401500250

CAS: 569-58-4 | C22H23N3O9 | 473.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 569-58-4
Molecular Formula C22H23N3O9
Molecular Weight (g/mol) 473.44
InChI Key AIPNSHNRCQOTRI-UHFFFAOYSA-N
Synonym aluminon, ammonium aurintricarboxylate, aurintricarboxylic acid ammonium salt, aurintricarboxylic acid, acs
PubChem CID 54729869
ChEBI CHEBI:87398
IUPAC Name triazanium;5-[(3-carboxy-4-oxidophenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-oxidobenzoate
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)[O-])C(=O)[O-])C(=O)O)[O-].[NH4+].[NH4+].[NH4+]

Specifications

Melting Point 220.0°C to 225.0°C
Ignition Residue 0.2% max
Packaging Glass bottle
Quantity 25 g
Beilstein 10,II,775
Merck Index 15,318
Solubility Information Solubility in water: 800g/L (20°C). Other solubilities: slightly soluble in ethanol,insoluble in petroleum ether
Formula Weight 473.44
Grade ACS Reagent
Insoluble Matter 0.1% max
Chemical Name or Material Aluminon
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiou
EINECSNumber 209-319-1

RUO – Research Use Only