missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate octadecahydrate, extra pure

Catalog No. p-45328
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123680010 1 kg Plastic bottle
AC123680025 2.5 kg Plastic bottle
AC123680100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC123680010 Supplier Thermo Scientific Chemicals Supplier No. 123680010
Only null left

CAS: 7784-31-8 | Al2O12S3·18H2O | 666.4 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2O12S3·18H2O
Molecular Weight (g/mol) 666.4
MDL Number MFCD00149136
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 86.0°C
Color Colorless to Yellow
pH 3 to 4 (2% soln. at 20°C)
Physical Form Liquid
Quantity 1 kg
Linear Formula Al2(SO4)3·18H2O
Merck Index 15,36
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 666.4
Percent Purity 98.0 to 110.0% (Complexometry)
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Aluminum sulfate octadecahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
RTECSNumber WS5697000

RUO – Research Use Only