Learn More
Aluminum sulfate octadecahydrate, 98+%, ACS reagent
CAS: 7784-31-8 | Al2O12S3·18H2O | 666.4 g/mol
Supplier: Thermo Scientific Chemicals 400590025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum sulfate octadecahydrate | |
| 7784-31-8 | |
| Crystalline Powder | |
| 98.0 to 102% (Complexometry) | |
| Al2(SO4)3·18H2O | |
| 15, 360 | |
| Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
| O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] | |
| 666.4 | |
| CHEBI:74779 | |
| ≥98% | |
| Plastic bottle |
| 86.0°C | |
| White | |
| 2.5 kg | |
| Al2O12S3·18H2O | |
| MFCD00149136 | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
| dialuminum;trisulfate;octadecahydrate | |
| 22377415 | |
| 666.4 | |
| ACS Reagent |
Chemical Identifiers
| 7784-31-8 | |
| 666.4 | |
| AMVQGJHFDJVOOB-UHFFFAOYSA-H | |
| 22377415 | |
| dialuminum;trisulfate;octadecahydrate |
| Al2O12S3·18H2O | |
| MFCD00149136 | |
| aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18 | |
| CHEBI:74779 | |
| O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3] |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
RTECSNumber : WS5697000
RUO – Research Use Only