missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate, pure, anhydrous

Catalog No. p-7077768
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458990010 1 kg
AC458990025 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458990010 Supplier Thermo Scientific Chemicals Supplier No. 458990010
Only null left

CAS: 10043-67-1 | AlKO8S2 | 258.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10043-67-1
Molecular Formula AlKO8S2
Molecular Weight (g/mol) 258.21
InChI Key GRLPQNLYRHEGIJ-UHFFFAOYSA-J
Synonym potassium alum, potash alum, aluminum potassium sulfate, potassium aluminum sulfate, burnt potassium alum, aluminum potassium disulfate, exsiccated alum, alum potassium, burnt alum, aluminum potassium alum
PubChem CID 24856
ChEBI CHEBI:86463
IUPAC Name aluminum;potassium;disulfate
SMILES [O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Specifications

Boiling Point 200.0°C
Melting Point 92.0°C
Quantity 1 kg
Solubility Information Solubility in water: 50g/L (20°C).
Formula Weight 258.21
Percent Purity ≥90% (Complexometry)
Grade Pure
Loss on Drying 1% max.
Chemical Name or Material Aluminum potassium sulfate
EINECSNumber 233-141-3

RUO – Research Use Only