missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum hydroxide, extra pure, powder
CAS: 21645-51-2 | AlH3O3 | 78.00 g/mol
Supplier: Thermo Scientific Chemicals 219130100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Aluminum hydroxide | |
| 21645-51-2 | |
| Powder | |
| AlH3O3 | |
| MFCD00003420 | |
| Solubility in water: insoluble | |
| [OH-].[OH-].[OH-].[Al+3] | |
| 78.00 | |
| ≥63.5% (Al2O3) | |
| 36.5% max. |
| White | |
| 8.5 to 10 (5% aq. suspension) | |
| 10 kg | |
| Al(OH)3 | |
| 15,338 | |
| WNROFYMDJYEPJX-UHFFFAOYSA-K | |
| aluminium(3+) trihydroxide | |
| 78 | |
| Extra Pure | |
| Plastic drum |
Chemical Identifiers
| 21645-51-2 | |
| 78.00 | |
| WNROFYMDJYEPJX-UHFFFAOYSA-K | |
| [OH-].[OH-].[OH-].[Al+3] |
| AlH3O3 | |
| MFCD00003420 | |
| aluminium(3+) trihydroxide |
Safety and Handling
EINECSNumber : 244-492-7
RUO – Research Use Only