missing translation for 'onlineSavingsMsg'
Learn More

Alternate Metals 3 For AA and ICP, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$580.44 - $1035.34

Specifications

Chemical Name or Material Alternate Metals Standard 3
Name Note For ICP/AA Alternate Water Pollution and Wastewater Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MN-4
View Documents
SPEX CertiPrep Group LLC
MN4
125 mL
Each for $580.44
Only null left
 
MN-4-500
View Documents
SPEX CertiPrep Group LLC
MN4500
500 mL
Each for $1,035.34
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca500μg/mL
Na500μg/mL
K100μg/mL
Mg100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Alternate Metals Standard 3
Triple-leached LDPE bottle
AA, ICP
Certified Reference Material
Environmental
For ICP/AA Alternate Water Pollution and Wastewater Standard
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
2% HNO3
Components in 2% HNO3:
Ca    •500μg/mL
Mg    •100μg/mL
K    •100μg/mL
Na    •500μg/mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product intended for laboratory use only