missing translation for 'onlineSavingsMsg'
Learn More

Alternate Metals 3 For AA and ICP, SPEX CertiPrep™

Catalog No. MN4500
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MN4500 500 mL
MN4 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MN4500 Supplier SPEX CertiPrep Group LLC Supplier No. MN4500
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca500μg/mL
Na500μg/mL
K100μg/mL
Mg100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Alternate Metals Standard 3
Packaging Triple-leached LDPE bottle
Name Note For ICP/AA Alternate Water Pollution and Wastewater Standard
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 500 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Ca    •500μg/mL
Mg    •100μg/mL
K    •100μg/mL
Na    •500μg/mL
Show More Show Less

Product intended for laboratory use only