missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTEX Standard Formulations, High Level, SPEX CertiPrep™
Description
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| Benzene | 71-43-2 | 2000μg/mL |
| Ethylbenzene | 100-41-4 | 2000μg/mL |
| m-Xylene | 108-38-3 | 2000μg/mL |
| o-Xylene | 95-47-6 | 2000μg/mL |
| p-Xylene | 106-42-3 | 2000μg/mL |
| Toluene | 108-88-3 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | BTEX Standard (High Level) |
| Packaging | 2mL Amber Prescored Glass Ampule |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity. |
| Methods | For US EPA Method 602 |
| Technique | AA, ICP |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Methanol-P and T |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 2000μg/mL |
Product intended for laboratory use only