missing translation for 'onlineSavingsMsg'
Learn More

Alternate Analytes B For CLP, SPEX CertiPrep™

Catalog No. INTB2
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
INTB2 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. INTB2 Supplier SPEX CertiPrep Group LLC Supplier No. INTB2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Al100μg/mL
As100μg/mL
B100μg/mL
Mo100μg/mL
Na100μg/mL
Sb100μg/mL
Se100μg/mL
Tl100μg/mL
Ca10μg/mL
Fe10μg/mL
Mg10μg/mL
Si10μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Alternate Interferents B
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique GC-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Concentration or Composition (by Analyte or Components) Components in 5% HNO3:
Al    •100Glc mug/mL
Sb    •100Glc mug/mL
As    •100Glc mug/mL
B    •100Glc mug/mL
Ca    •10Glc mug/mL
Fe    •10Glc mug/mL
Mg    •10Glc mug/mL
Mo    •100Glc mug/mL
Se    •100Glc mug/mL
Si    •10Glc mug/mL
Na    •100Glc mug/mL
Tl    •100Glc mug/mL

Product intended for laboratory use only