missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559252
Description
Immunogen sequence: DCGCPLKALR DFALQNPSAV PRFVQAICEG DDCQPPAYTY NNITCASPPE VVGLDLRDLS EAHFAP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| ALS | |
| Polyclonal | |
| Unconjugated | |
| IGFALS | |
| ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P35858 | |
| IGFALS | |
| Recombinant protein corresponding to Human ALS. Recombinant protein control fragment (Product #RP-98357). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |