Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
Catalog No. AB1118
Change view
Click to view available options
Description:
384-well PCR Plate Carrier
96-Well PCR Plate Carrier
Standard Microplate Carrier
V Bottom 96-Well Storage Plate Carrier
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
AB1118 V Bottom 96-Well Storage Plate Carrier
AB05631000 96-Well PCR Plate Carrier
AB0724 384-well PCR Plate Carrier
AB0827 Standard Microplate Carrier
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AB1118 Supplier Thermo Scientific™ Supplier No. AB1118
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 year
Specifications
Description V Bottom 96-Well Storage Plate Carrier
For Use With (Equipment) ALPS™ Heat Sealer
No. per Pack 1 Ea.
Product Type Carrier