Learn More
Thermo Scientific™ ALPS Heat Sealer Accessories
Use these ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer.
Supplier: Thermo Scientific™ AB0827
Description
Use Thermo Scientific™ ALPS heat sealer accessories with the ALPS 25 Manual Heat Sealer and ALPS 50 V Microplate Heat Sealer. The foil stripper, AB-0592, is also compatible with the ALPS 3000 Microplate Heat Sealer.
Warranty: 1 yearChemical Identifiers
| ALPS™ Heat Sealer |
Specifications
| Standard Microplate Carrier | |
| ALPS™ Heat Sealer | |
| Carrier |