missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALPL Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566055
Description
ALPL Monoclonal Antibody for IHC (P), ICC/IF
Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism's environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.Specifications
| ALPL | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P05186 | |
| ALPL | |
| Recombinant Alkaline Phosphatase, Tissue-nonspecific (ALPL) (Leu18-Arg335). | |
| 200 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| C1 | |
| Unconjugated | |
| ALPL | |
| Akp2; Akp-2; Alkaline phosphatase 2; alkaline phosphatase 2, liver; alkaline phosphatase liver/bone/kidney isozyme; alkaline phosphatase precursor (EC 3.1.3.2); alkaline phosphatase tissue non-specific isoform; alkaline phosphatase tissue non-specific isoform/TNS-AP; alkaline phosphatase, biomineralization associated; alkaline phosphatase, intestinal; alkaline phosphatase, liver/bone/kidney; alkaline phosphatase, tissue-nonspecific isozyme; alkaline phosphomonoesterase; ALPI; Alpl; APTNAP; AP-TNAP; FLJ40094; FLJ93059; glycerophosphatase; HOPS; IAP; intestinal alkaline phosphatase; intestinal-type alkaline phosphatase; Kasahara isozyme; liver/bone/kidney isozyme; liver/bone/kidney-type alkaline phosphatase; MGC161443; MGC167935; PHOA; tissue-nonspecific alkaline phosphatase; tissue-nonspecific alkaline phosphatase precursor; tissue-nonspecific ALP; Tissue-nonspecific ALP alkaline phosphatase; TNAP; TNSALP; TNS-AP | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100170147, 249 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |