missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

Supplier:  Ansell 3718511080

 View more versions of this product

Catalog No. 11-391-140B



Only null left
Explore available promotions

Description

Description

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

  • High performance nitrile compound
  • Combination of chemical resistance and strength ,for optimal results in wet or dry environments
  • Snag, puncture and abrasion protection compared with rubber or neoprene gloves
  • High levels of flexibility, comfort and dexterity
  • Suitable for heavy-duty cleaning applications
  • Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications

Specifications

ANSI 4
CE 0493, EN 420:2003 + A1:2009, Category III
Industrial
Yes
Chemical-resistant Gloves
22 mil
EN 388:4102X, EN 388:4102
Type A
Bacteria, Fungi only
Gauntlet
Unlined
37-185 8 117299
18 in.
FDA 21 CFR 177.2600
Yes
Green
Nitrile
8
0.55 mm
No
AGJKLPT
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Sandpatch
ISO 18889
AlphaTec™ Solvex™ 37-185
45.7 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions