missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Alphatec™ MICROCHEM™ 68-4000 Green Coveralls with Hood and Socks
Provide an exceptional barrier against organic and inorganic chemials as well as biological agents. Ansell™ Alphatec™ MICROCHEM™ 68-4000 Green Coveralls with Hood and Socks incorporate unique design features that enhance wearer comfort and safety.
Supplier: Ansell™ GR40T9212205
Alert:
This product contains natural rubber latex which may cause allergic reactions.
Description
- Protection: Permeation tested against over 190 chemicals, including chemical warfare agents; fully encapsulated versions also available
- Comfort: Textile like interior improves wearer acceptance
- Anti-static: Tested according to EN1149-5 & AATCC76 (8EMSL test method)
- Design: Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece respirator fit hood, integrated socks with boot overflap
- Applications: Chemicals handling/transportation, oil-based mud protection, hazardous waste remediation, sewage purification installations, indsutrial/tank cleaning, HAZMAT emergency response (Level B), pharmaceutical, mining, agriculture
- Sizes: Available in Small through 5X-Large
- Certifications: EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
- Storage: 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
Specifications
| X-Large | |
| Green | |
| Attached Socks, Boot Overflaps | |
| Hooded | |
| Double Zipper | |
| Liquid and spray tight chemical and biological protection | |
| No |
| Multi-layer Barrier Laminate Fabric | |
| Elastic | |
| Elastic, Knit | |
| 2-panel, Respirator Fit | |
| Welded, Taped | |
| Elasticated outer cuff with knitted inner cuff |