missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Alphatec™ MICROCHEM™ 68-4000 Green Coveralls with Hood and Socks

Catalog No. p-7070759
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18000012 Small
18000013 Medium
18000014 Large
18000015 X-Large
18000016 2X-Large
18000017 3X-Large
18000018 4X-Large
18000019 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 18000012 Supplier Ansell™ Supplier No. GR40T9212202
Only null left
This product contains natural rubber latex which may cause allergic reactions.

Provide an exceptional barrier against organic and inorganic chemials as well as biological agents. Ansell™ Alphatec™ MICROCHEM™ 68-4000 Green Coveralls with Hood and Socks incorporate unique design features that enhance wearer comfort and safety.

  • Protection:  Permeation tested against over 190 chemicals, including chemical warfare agents; fully encapsulated versions also available
  • Comfort:  Textile like interior improves wearer acceptance
  • Anti-static:  Tested according to EN1149-5 & AATCC76 (8EMSL test method)
  • Design:  Elastic waist, wrists and ankles, double zip closure, double cuffs with knitted inner cuff, ultrasonically welded and taped seams, 2 piece respirator fit hood, integrated socks with boot overflap
  • Applications:  Chemicals handling/transportation, oil-based mud protection, hazardous waste remediation, sewage purification installations, indsutrial/tank cleaning, HAZMAT emergency response (Level B), pharmaceutical, mining, agriculture
  • Sizes:  Available in Small through 5X-Large
  • Certifications:  EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B
  • Storage:  5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5-B, ISO 17491-4 Type 6-B, EN 14126 / ASTM F1671, EN 14605 Type 3-B, EN 14605 Type 4-B

Specifications

Size Small
Material Multi-layer Barrier Laminate Fabric
Color Green
Waist Style Elastic
Ankle Style Attached Socks, Boot Overflaps
Cuff Style Elastic, Knit
Neck Style Hooded
Hood Type 2-panel, Respirator Fit
Closure Type Double Zipper
Seam Type Welded, Taped
Protection Class Liquid and spray tight chemical and biological protection
Description Elasticated outer cuff with knitted inner cuff
Latex Free No
Show More Show Less