Learn More
Ansell AlphaTec™ 2000 Comfort Bound Coveralls with Hood, Model 129
Coveralls protective barrier shields hood, arms, legs, and front torso against particulates, liquids, and light chemical splash. Three-piece hood ensures adequate coverage and finger loops improve comfort and usability. Previously known as MICROCHEM™ by AlphaTec 2000 Comfort.
Supplier:  Ansell WH20B9212904
 View more versions of this product
Catalog No. 17-999-886
Only null left
Explore available promotions
Description
Description
  • Arms, legs, and front torso in 2000 fabric
  • Protects against liquid, light chemical resistance,and particulate biological agents
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free
  • Anti-static, tested according to EN1149-5 and AATCC76
  • Elastic ankles and wrists
  • 3-piece hood, 2-way front zipper with storm flap
  • Bound seams, finger loops
  • Ideal for pharmaceutical industries, cleanrooms, paint spraying, veterinary services, pest control
  • Available in Small through 5X-Large
  • Certifications EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 8
Specifications
Specifications
General Purpose Coveralls
Unisex
White
Elastic
Elastic, Finger Loops
3-panel, Respirator Fit
Bound
Non-toxic Particulates, Liquid, Chemicals
AATTC 76 Compliant
Yes
Automotive, Life Sciences, Metal Fabrication, Veterinary, Maintenance, Cleaning, Production, Food Processing
Large
Microporous Laminate, SMS
Limited-use
Elastic
Hooded
Front Zipper with Storm Flap
Non-sterile
Breathable SMS Back Panel
EN 1149-5, CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
WH20-B-92-129
Product Suggestions
Product Suggestions