missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 2000 Comfort Bound Coveralls with Hood, Model 129

Catalog No. p-7070777
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999884 Small
17999886 Large
17999885 Medium
17999887 X-Large
17999888 2X-Large
17999889 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17999884 Supplier Ansell Supplier No. WH20B9212902
Only null left

Coveralls protective barrier shields hood, arms, legs, and front torso against particulates, liquids, and light chemical splash. Three-piece hood ensures adequate coverage and finger loops improve comfort and usability. Previously known as MICROCHEM™ by AlphaTec 2000 Comfort.

  • Arms, legs, and front torso in 2000 fabric
  • Protects against liquid, light chemical resistance,and particulate biological agents
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone free
  • Anti-static, tested according to EN1149-5 and AATCC76
  • Elastic ankles and wrists
  • 3-piece hood, 2-way front zipper with storm flap
  • Bound seams, finger loops
  • Ideal for pharmaceutical industries, cleanrooms, paint spraying, veterinary services, pest control
  • Available in Small through 5X-Large
  • Certifications EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1073-2, EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

Specifications

Product Type General Purpose Coveralls
Size Small
Gender Unisex
Material Microporous Laminate, SMS
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Respirator Fit
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Chemicals
Back Style Breathable SMS Back Panel
Certifications/Compliance AATTC 76 Compliant
Certifications/Compliance (Europe) EN 1149-5, CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN 1149-5)
Latex Free Yes
Model WH20-B-92-129
For Use With (Application) Automotive, Life Sciences, Metal Fabrication, Veterinary, Maintenance, Cleaning, Production, Food Processing
Show More Show Less