missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Alphatec™ 1800 Standard Bound Model 103 Coveralls with Collar
Coveralls with collar provide entry level, low hazard liquid and particulate protection for a wide range of general industrial applications. Previously known as MICROCHEM™ by AlphaTec 1800.
Supplier: Ansell WH18B9210307
Alert:
This product contains natural rubber latex which may cause allergic reactions.
Description
- Provides low hazard liquid spray, splash and particle protection
- Moisture vapor permeable to help reduce the risk of heat stress
- Low linting reduces the risk of fiber contamination in some critical areas
- Anti-static, tested according to EN1149-5 and AATCC76 (8EMSL test method)
- Elastic waist, wrists, and ankles
- 2-way front zipper with storm flap
- Bound seams, collar
- Available in Small through 5X-Large sizes
- Certifications EN 1149-5, AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
- 5 years shelf life in dry, cool, covered conditions out of direct sunlight
Certifications
EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 11
Specifications
| General Purpose Coveralls | |
| Unisex | |
| White | |
| Elastic | |
| Elastic | |
| Front Zipper with Storm Flap | |
| Non-sterile | |
| AATCC 76, ANSI/ISEA 101 | |
| No | |
| None |
| 3X-Large | |
| Microporous Film Laminate | |
| Limited-use | |
| Elastic | |
| Collared | |
| Bound | |
| Non-toxic Particulates, Liquid, Anti-static | |
| EN 1149-5, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6 | |
| WH18-B-92-103 | |
| General Maintenance, Paint Spraying, Powder Handling, Food |