missing translation for 'onlineSavingsMsg'
Learn More

Ansell Alphatec™ 1800 Standard Bound Model 103 Coveralls with Collar

Catalog No. p-7070771
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17999965 Medium
17999964 Small
17999966 Large
17999967 X-Large
17999968 2X-Large
17999969 3X-Large
17999970 4X-Large
17999971 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This product contains natural rubber latex which may cause allergic reactions.
Catalog No. 17999965 Supplier Ansell Supplier No. WH18B9210303
Only null left
This product contains natural rubber latex which may cause allergic reactions.

Coveralls with collar provide entry level, low hazard liquid and particulate protection for a wide range of general industrial applications. Previously known as MICROCHEM™ by AlphaTec 1800.

  • Provides low hazard liquid spray, splash and particle protection
  • Moisture vapor permeable to help reduce the risk of heat stress
  • Low linting reduces the risk of fiber contamination in some critical areas
  • Anti-static, tested according to EN1149-5 and AATCC76 (8EMSL test method)
  • Elastic waist, wrists, and ankles
  • 2-way front zipper with storm flap
  • Bound seams, collar
  • Available in Small through 5X-Large sizes
  • Certifications EN 1149-5, AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
  • 5 years shelf life in dry, cool, covered conditions out of direct sunlight

Certifications

EN 1149-5 / AATCC 76, ANSI/ISEA 101, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Non-toxic Particulates, Liquid, Anti-static
Certifications/Compliance AATCC 76, ANSI/ISEA 101
Certifications/Compliance (Europe) EN 1149-5, EN ISO 13982-1 Type 5, ISO 17491-4 Type 6
Latex Free No
Model WH18-B-92-103
No. of Pockets None
For Use With (Application) General Maintenance, Paint Spraying, Powder Handling, Food
Show More Show Less