missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 87-320 Rubber Gloves
Flocked-lined natural rubber gloves, combining comfort, flexibility and food compliance.
$254.07 - $254.07
Specifications
| Certifications/Compliance | EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, Category III |
|---|---|
| Certifications/Compliance (Europe) | CE 0493, EN 388:2016 +A1:2018, REACH Compliant |
| Chlorinated | No |
| Applications | Industrial |
| Color | Orange |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
17-609-976
|
Ansell
87320070* |
7 |
Case of 144 Pair for $254.07
|
|
|||||
|
17-609-977
|
Ansell
87320080* |
8 |
Case of 144 Pair for $254.07
|
|
|||||
|
17-609-978
|
Ansell
87320090* |
9 |
Case of 144 Pair for $254.07
|
|
|||||
|
17-609-979
|
Ansell
87320100* |
10 |
Case of 144 Pair for $254.07
|
|
|||||
Description
Key Features and Benefits
- Beaded cuff: For easy-to-wear chemical gloves with tear resistance
- Food-safe: Excellent hand and food safety gloves
- Good abrasion resistance: Protective gloves for use in low-hazard areas
Description
- Increased protection and convenience: Flock-lined and lightweight, AlphaTec™ 87-320 rubber work gloves come with beaded cuffs, which ensure tear resistance and easy donning and doffing
- Food handling compliance: Featuring an AQL of 1.5, these orange food handling gloves are suitable for a wide range of food applications
- Enhanced durability: AlphaTec™ 87-320 hand protection gloves are also abrasion-resistant, lending themselves to low-hazard tasks
- Increased comfort: Owing to their 100% cotton flock lining, these flexible work gloves are comfortable and offer wearers the required flexibility for light-duty tasks
Specifications
| EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, Food Contact, Category III | |
| No | |
| Orange | |
| 0.45 mm | |
| Rubber Latex | |
| 0.45 mm | |
| 18 mil | |
| EN 388:2016 +A1:2018 - 2010X | |
| Type A | |
| Bacteria, Fungi WITH Virus Protection | |
| Raised Diamond | |
| Flock Lined | |
| 300 mm |
| CE 0493, EN 388:2016 +A1:2018, REACH Compliant | |
| Industrial | |
| 18 mil | |
| No | |
| 18 mil | |
| Chemical Protection Gloves | |
| 0.45 mm | |
| No | |
| KLT | |
| Beaded | |
| Cotton Flock | |
| 11.8 in. |