missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-535B Chemical Resistant Nitrile Gloves With ANSELL GRIP™

Teal and black insulated nitrile gloves feature a combination of chemical resistance, confident grip and comfortable inner lining.

Supplier:  Ansell 58535B080

 View more versions of this product

Catalog No. 17-999-827



Only null left
Explore available promotions

Description

Description

  • ANSELL GRIP™ technology to handle wet or oily objects with less grip force and more control
  • Combination of liquid-proof chemical resistance and grip
  • Coating is consistent to reduce the likelihood of chemical leakage
  • Recommended for outdoor applications
  • Black acrylic liner
Specifications

Specifications

ANSI 5
CE 0493, EN 420:2003 + A1:2009, Category III, EN 1149, EN 407
Gray, Teal
Nitrile
12 mil
EN 388:3122A, EN 388:4121
Type A
Bacteria, Fungi only
ANSELL GRIP™ Technology
Lined
AlphaTec™ 58-535B
34 cm
21 CFR 170-199
Yes
Yes
8
0.30 mm
No
AJKLOPT
Gauntlet
Black Acrylic
58535B080
13.3 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions