missing translation for 'onlineSavingsMsg'
Learn More

HER6 Antibody, ProSci

Catalog No. 16824800
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16824800 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16824800 Supplier ProSci Llc Supplier No. 457260100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Goat Polyclonal Antibody

HER6 Polyclonal Antibody specifically detects HER6. It is validated for ELISA

Specifications

Antigen HER6
Applications ELISA
Classification Polyclonal
Concentration 500 ug/mL
Conjugate Unconjugated
Dilution Peptide ELISA: antibody detection limit dilution 1:128000.Western Blot:Not yet tested. At this stage we are dependent on researchers in the field for further characterization of this product. Therefore we cannot recommend an optimal concentration and the product is investigative grade. We would appreciate any feedback from people in the field - have any results been reported with other antibodies/lysates?
Formulation Supplied at 0.5 mg/mL in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin. Aliquot and store at -20°C. Minimize freezing and thawing.
Gene Accession No. NP_571154.1
Gene Alias her6, hairy-related 6 , HER-6, cb129
Host Species Goat
Purification Method Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30288
Target Species Not Available
Content And Storage Aliquot and store at -20°C. Minimize freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Cancer
Show More Show Less